Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600002936.mm.1 |
TC0600000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002936.mm.1 |
NM_001170848.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002936.mm.1 |
NM_001170849.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002936.mm.1 |
NM_138680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002944.mm.1 |
TC0600000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002944.mm.1 |
NM_001170848.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002944.mm.1 |
NM_138680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002945.mm.1 |
TC0600000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002945.mm.1 |
NM_001170848.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002945.mm.1 |
NM_138680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002947.mm.1 |
TC0600000381.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002947.mm.1 |
NM_001170848.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002947.mm.1 |
NM_138680.3 |
Refseq |