Assay Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011539.3 | NP_035669.3 | 11 - 12 | 1537 | 65 | |
| GenBank mRNA | BC029014.1 | - | 11 - 12 | 1539 | 5007629 | 65 |
| L18868.1 | - | 11 - 12 | 1408 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbxas1
Genomic Map