Assay Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011539.3 | NP_035669.3 | 2 - 3 | 348 | 76 | |
| GenBank mRNA | AK048544.1 | - | 2 - 3 | 330 | 76 | |
| AK139479.1 | - | 2 - 3 | 311 | 76 | ||
| AK162394.1 | - | 453 | 76 | |||
| BC029014.1 | - | 2 - 3 | 350 | 5007629 | 76 | |
| L18868.1 | - | 2 - 3 | 219 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbxas1
Genomic Map