Assay Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013792.2 | NP_001013814.2 | 5 - 6 | 684 | 64 | |
| GenBank mRNA | AK134955.1 | - | 5 - 6 | 519 | 64 | |
| AK150371.1 | - | 5 - 6 | 687 | 64 | ||
| BC087945.1 | - | 2 - 3 | 285 | 30505408 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otulin
Genomic Map