Assay Details
| Gene Symbol: Ank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020332.4 | NP_065065.3 | 6 - 7 | 1152 | 99 | |
| GenBank mRNA | AF001533.2 | - | 4 - 5 | 498 | 99 | |
| AF274752.1 | - | 6 - 7 | 855 | 99 | ||
| AK083135.1 | - | 6 - 7 | 1153 | 99 | ||
| AK148070.1 | - | 6 - 7 | 925 | 99 | ||
| BC042644.1 | - | 6 - 7 | 1077 | 99 | ||
| BC054379.1 | - | 6 - 7 | 1059 | 6390764 | 99 | |
| DQ832285.1 | - | 6 - 7 | 1152 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ank
Genomic Map