Assay Details
| Gene Symbol: Hinfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172162.3 | NP_751894.2 | 5 - 6 | 729 | 92 | |
| GenBank mRNA | AK049348.1 | - | 5 - 6 | 691 | 92 | |
| AK049633.1 | - | 5 - 6 | 890 | 92 | ||
| BC029091.1 | - | 4 - 5 | 712 | 2651006 | 92 | |
| CT010239.1 | - | 4 - 5 | 664 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hinfp
Genomic Map