Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017913.mm.1 |
NM_027909.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017913.mm.1 |
NM_027909.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017913.mm.1 |
TC0900002156.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017916.mm.1 |
NM_027909.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017916.mm.1 |
NM_027909.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017916.mm.1 |
TC0900002156.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017914.mm.1 |
NM_027909.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017914.mm.1 |
NM_027909.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017914.mm.1 |
TC0900002156.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017915.mm.1 |
NM_027909.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017915.mm.1 |
NM_027909.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017915.mm.1 |
TC0900002156.mm.1 |
MTA TC |