Assay Details
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007619.2 | NP_031645.2 | 11 - 12 | 2090 | 72 | |
| GenBank mRNA | AK029826.1 | - | 11 - 12 | 2093 | 72 | |
| AK153915.1 | - | 11 - 12 | 2042 | 72 | ||
| AK163907.1 | - | 10 - 11 | 1916 | 72 | ||
| BC125285.1 | - | 11 - 12 | 2005 | 40129800 | 72 | |
| X57111.1 | - | 11 - 12 | 1977 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbl
Genomic Map