Assay Details
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052994.2 | NP_443720.1 | 10 - 11 | 1338 | 66 | |
GenBank mRNA | AJ278999.1 | - | 10 - 11 | 1340 | 66 | |
AK122233.1 | - | 10 - 11 | 1000 | 66 | ||
AK140483.1 | - | 10 - 11 | 1245 | 66 | ||
BC057324.1 | - | 10 - 11 | 1338 | 6821449 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P221756 | BC057324.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P221756 | AK122233.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P221756 | NM_052994.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P221756 | BC057324.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P221756 | AK122233.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P221756 | NM_052994.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29445 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742361 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319860 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439400 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353186 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598130 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8607 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265168 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703858 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512156 | BC027186.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180062 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99572 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156070 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118797 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153619 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P221756 | BC057324.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P221756 | AK122233.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P221756 | NM_052994.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P221756 | BC057324.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P221756 | AK122233.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P221756 | NM_052994.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07074409 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02135474 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02135486 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153619 | - | - |
Whole Mouse Genome G4122A | A_52_P385801 | BC060210.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P385801 | AK122233.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P221756 | BC057324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P221756 | AK122233.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P221756 | NM_052994.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P221756 | BC057324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P221756 | AK122233.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P221756 | NM_052994.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P393181 | BC057324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P393181 | BC060210.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P393181 | AJ278999.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P393181 | AK122233.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P393181 | AK140483.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117698 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121494 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02135474 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153619 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08890339 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08397289 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08890341 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08890336 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08397296 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08397292 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08397288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27172577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08322698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08322706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08322700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08791852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08781433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27172681 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10363425 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363418 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363423 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363419 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363416 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363426 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363417 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363428 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363422 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363421 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363420 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363427 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10363424 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435026_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450451_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10363415 | - | - |
Mouse Genome 430 2.0 Array | 1458360_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450451_at | - | - |
Mouse Genome 430 2.0 Array | 1458360_at | - | - |
Mouse Genome 430 2.0 Array | 1457430_at | - | - |
Mouse Genome 430 2.0 Array | 1457430_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435026_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435026_at | - | - |
Mouse Genome 430 2.0 Array | 1447443_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450451_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447443_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94214_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94214_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000003673.mm.1 | NM_052994.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000003673.mm.1 | NM_052994.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000003673.mm.1 | TC1000000543.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000003674.mm.1 | NM_052994.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000003674.mm.1 | NM_052994.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000003674.mm.1 | TC1000000543.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000003675.mm.1 | NM_052994.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000003675.mm.1 | NM_052994.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000003675.mm.1 | TC1000000543.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435026_at | - | - |
Mouse Expression Set 430 | 1435026_PM_at | - | - |
Mouse Expression Set 430 | 1450451_PM_at | - | - |
Mouse Expression Set 430 | 1450451_at | - | - |
Murine Genome U74v2 Set | 104375_PM_at | - | - |
Murine Genome U74v2 Set | 102095_f_at | - | - |
Murine Genome U74v2 Set | 104375_at | - | - |
Murine Genome U74v2 Set | 102095_PM_f_at | - | - |
Murine Genome U74v2 Set | 137714_at | - | - |
Murine Genome U74v2 Set | 137714_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435026_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435026_at | - | - |
Mouse Genome 430A 2.0 Array | 1450451_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450451_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435026_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1435026_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450451_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450451_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6650369 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6650369 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006650369 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251096 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251096 | - | - |
mouse-6 v1.1 (Illumina) | scl094214.11_38-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6650369 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl094214.11_38-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650369 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1251096 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251096 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl094214.11_38-S | - | - |
