Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000003658.mm.1 |
NM_052994.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003658.mm.1 |
NM_052994.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003658.mm.1 |
TC1000000543.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003663.mm.1 |
NM_052994.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003663.mm.1 |
NM_052994.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003663.mm.1 |
TC1000000543.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003660.mm.1 |
NM_052994.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003660.mm.1 |
NM_052994.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003660.mm.1 |
TC1000000543.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003659.mm.1 |
NM_052994.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003659.mm.1 |
NM_052994.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003659.mm.1 |
TC1000000543.mm.1 |
MTA TC |