Assay Details
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052994.2 | NP_443720.1 | 3 - 4 | 451 | 84 | |
GenBank mRNA | AJ278999.1 | - | 3 - 4 | 453 | 84 | |
AK122233.1 | - | 3 - 4 | 113 | 84 | ||
AK140483.1 | - | 3 - 4 | 357 | 84 | ||
AK162941.1 | - | 3 - 4 | 356 | 84 | ||
BC057324.1 | - | 3 - 4 | 451 | 6821449 | 84 | |
BC060210.1 | - | 3 - 4 | 403 | 6838067 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
