Assay Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027425.3 | NP_081701.2 | 11 - 12 | 1214 | 108 | |
| GenBank mRNA | AK083205.1 | - | 11 - 12 | 1199 | 108 | |
| AK173195.1 | - | 11 - 12 | 1175 | 108 | ||
| BC137736.1 | - | 11 - 12 | 1163 | 8860755 | 108 | |
| BC137737.1 | - | 11 - 12 | 1163 | 8860756 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rufy2
Genomic Map