Assay Details
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027425.3 | NP_081701.2 | 8 - 9 | 832 | 108 | |
GenBank mRNA | AK011847.1 | - | 8 - 9 | 802 | 108 | |
AK083205.1 | - | 8 - 9 | 817 | 108 | ||
AK173195.1 | - | 8 - 9 | 793 | 108 | ||
BC017645.1 | - | 8 - 9 | 794 | 108 | ||
BC137736.1 | - | 8 - 9 | 781 | 8860755 | 108 | |
BC137737.1 | - | 8 - 9 | 781 | 8860756 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
