Assay Details
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027425.3 | NP_081701.2 | 9 - 10 | 925 | 79 | |
GenBank mRNA | AK011847.1 | - | 9 - 10 | 895 | 79 | |
AK083205.1 | - | 9 - 10 | 910 | 79 | ||
AK173195.1 | - | 9 - 10 | 886 | 79 | ||
BC017645.1 | - | 9 - 10 | 887 | 79 | ||
BC038265.1 | - | 8 - 9 | 796 | 79 | ||
BC137736.1 | - | 9 - 10 | 874 | 8860755 | 79 | |
BC137737.1 | - | 9 - 10 | 874 | 8860756 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
