Assay Details
| Gene Symbol: Supv3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181423.2 | NP_852088.1 | 3 - 4 | 465 | 157 | |
| GenBank mRNA | AJ968954.1 | - | 3 - 4 | 492 | 157 | |
| AK031911.1 | - | 3 - 4 | 456 | 157 | ||
| AK143755.1 | - | 3 - 4 | 490 | 157 | ||
| BC049796.1 | - | 3 - 4 | 465 | 6332552 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supv3l1
Genomic Map