Assay Details
| Gene Symbol: Dna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177372.3 | NP_796346.2 | 13 - 14 | 2056 | 58 | |
| GenBank mRNA | AK028381.1 | - | 13 - 14 | 2033 | 58 | |
| AK129051.1 | - | 13 - 14 | 2041 | 58 | ||
| BC025182.1 | - | 8 - 9 | 1207 | 58 | ||
| BC030382.1 | - | 13 - 14 | 1997 | 58 | ||
| BC115716.1 | - | 13 - 14 | 2010 | 40085888 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dna2
Genomic Map