Assay Details
| Gene Symbol: Dna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177372.3 | NP_796346.2 | 6 - 7 | 1010 | 68 | |
| GenBank mRNA | AK028381.1 | - | 6 - 7 | 987 | 68 | |
| AK129051.1 | - | 6 - 7 | 995 | 68 | ||
| BC025182.1 | - | 1 - 2 | 161 | 68 | ||
| BC030382.1 | - | 6 - 7 | 952 | 68 | ||
| BC115716.1 | - | 6 - 7 | 964 | 40085888 | 68 | |
| CO433698.1 | - | 4 - 5 | 581 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dna2
Genomic Map