Assay Details
| Gene Symbol: Dna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177372.3 | NP_796346.2 | 8 - 9 | 1283 | 76 | |
| GenBank mRNA | AK028381.1 | - | 8 - 9 | 1260 | 76 | |
| AK129051.1 | - | 8 - 9 | 1268 | 76 | ||
| BC030382.1 | - | 8 - 9 | 1224 | 76 | ||
| BC115716.1 | - | 8 - 9 | 1237 | 40085888 | 76 | |
More Information
Set Membership: |
|

Dna2
Genomic Map