Assay Details
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282992.1 | NP_001269921.1 | 38 - 39 | 8609 | 63 | |
| NM_008787.3 | NP_032813.3 | 38 - 39 | 8663 | 63 | ||
| GenBank mRNA | AB207233.1 | - | 38 - 39 | 8483 | 63 | |
| AB207234.1 | - | 26 - 27 | 5904 | 63 | ||
| AK029906.1 | - | 16 - 17 | 4159 | 63 | ||
| AK122275.2 | - | 23 - 24 | 5356 | 63 | ||
| BC039661.1 | - | 4 - 5 | 626 | 63 | ||
| BC055467.1 | - | 5 - 6 | 710 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcnt
Genomic Map