Assay Details
| Gene Symbol: Ube2cbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027394.2 | NP_081670.1 | 8 - 9 | 1042 | 70 | |
| GenBank mRNA | AK011446.1 | - | 8 - 9 | 1059 | 70 | |
| AK049229.1 | - | 8 - 9 | 1043 | 70 | ||
| BC115686.1 | - | 8 - 9 | 1011 | 40104268 | 70 | |
| BC115687.1 | - | 8 - 9 | 1010 | 40104269 | 70 | |
More Information
Set Membership: |
|

Ube2cbp
Genomic Map