Assay Details
| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172920.4 | NP_766508.3 | 18 - 19 | 1693 | 71 | |
| GenBank mRNA | AK143845.1 | - | 19 - 20 | 1698 | 71 | |
| BC031382.1 | - | 11 - 12 | 871 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l1
Genomic Map