Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900016147.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016147.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016147.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016145.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016145.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016145.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016146.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016146.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016146.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016148.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016148.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016148.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016149.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016149.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016149.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016151.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016151.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016151.mm.1 |
TC0900001921.mm.1 |
MTA TC |