Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900016135.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016135.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016135.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016139.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016139.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016139.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016136.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016136.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016136.mm.1 |
TC0900001921.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016138.mm.1 |
NM_172920.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016138.mm.1 |
NM_172920.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016138.mm.1 |
TC0900001921.mm.1 |
MTA TC |