Assay Details
| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113323.1 | NP_001106794.1 | 15 - 16 | 1711 | 119 | |
| GenBank mRNA | AK014852.1 | - | 15 - 16 | 1544 | 119 | |
| BC132200.1 | - | 15 - 16 | 1504 | 40130477 | 119 | |
| BC132202.1 | - | 15 - 16 | 1504 | 40130479 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l3
Genomic Map