Assay Details
| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113323.1 | NP_001106794.1 | 1 - 2 | 265 | 76 | |
| GenBank mRNA | BC132200.1 | - | 1 - 2 | 58 | 40130477 | 76 |
| BC132202.1 | - | 1 - 2 | 58 | 40130479 | 76 | |
| DV051729.1 | - | 1 - 2 | 285 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC132200.1, BC132202.1, NM_001113323.1 transcripts.
Set Membership: |
|

Glb1l3
Genomic Map