Assay Details
| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113323.1 | NP_001106794.1 | 2 - 3 | 362 | 104 | |
| GenBank mRNA | BC132200.1 | - | 2 - 3 | 155 | 40130477 | 104 |
| BC132202.1 | - | 2 - 3 | 155 | 40130479 | 104 | |
| DV051729.1 | - | 2 - 3 | 382 | 104 | ||
More Information
Set Membership: |
|

Glb1l3
Genomic Map