Assay Details
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161623.1 | NP_001155095.1 | 2 - 3 | 187 | 62 | |
NM_009862.2 | NP_033992.2 | 3 - 4 | 426 | 62 | ||
GenBank mRNA | AF081536.1 | - | 3 - 4 | 426 | 62 | |
AF081537.1 | - | 2 - 3 | 125 | 62 | ||
AF081538.1 | - | 2 - 3 | 125 | 62 | ||
AF098068.1 | - | 2 - 3 | 172 | 62 | ||
AJ223729.1 | - | 2 - 3 | 143 | 62 | ||
AK147013.1 | - | 2 - 3 | 143 | 62 | ||
AK167850.1 | - | 2 - 3 | 175 | 62 | ||
BC006856.1 | - | 2 - 3 | 143 | 62 | ||
BC028635.1 | - | 2 - 3 | 179 | 1262927 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
