Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
TC0700002644.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
NM_001190974.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
NM_001190974.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
NM_001190975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
NM_001190975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
NM_009465.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022909.mm.1 |
NM_009465.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
TC0700002644.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
NM_001190974.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
NM_001190974.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
NM_001190975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
NM_001190975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
NM_009465.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022910.mm.1 |
NM_009465.4 |
Refseq |