Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004562.mm.1 |
NM_001145884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004562.mm.1 |
NM_001145884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004562.mm.1 |
NM_010580.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004562.mm.1 |
NM_010580.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004562.mm.1 |
TC1600000480.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004567.mm.1 |
NM_001145884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004567.mm.1 |
NM_001145884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004567.mm.1 |
NM_010580.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004567.mm.1 |
NM_010580.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004567.mm.1 |
TC1600000480.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004563.mm.1 |
NM_001145884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004563.mm.1 |
NM_001145884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004563.mm.1 |
NM_010580.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004563.mm.1 |
NM_010580.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004563.mm.1 |
TC1600000480.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004565.mm.1 |
NM_001145884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004565.mm.1 |
NM_001145884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004565.mm.1 |
NM_010580.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004565.mm.1 |
NM_010580.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004565.mm.1 |
TC1600000480.mm.1 |
MTA TC |