Assay Details
| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009655.2 | NP_033785.1 | 10 - 11 | 1769 | 125 | |
| GenBank mRNA | AB003026.1 | - | 10 - 11 | 1431 | 125 | |
| AK030851.1 | - | 10 - 11 | 1770 | 125 | ||
| AK031391.1 | - | 2845 | 125 | |||
| BC027280.1 | - | 10 - 11 | 3501215 | 125 | ||
| L25274.1 | - | 5 - 6 | 561 | 125 | ||
| U95030.1 | - | 10 - 11 | 1237 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alcam
Genomic Map