Assay Details
| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009655.2 | NP_033785.1 | 3 - 4 | 925 | 99 | |
| GenBank mRNA | AB003026.1 | - | 3 - 4 | 587 | 99 | |
| AK030851.1 | - | 3 - 4 | 926 | 99 | ||
| AK031391.1 | - | 2001 | 99 | |||
| BC027280.1 | - | 3 - 4 | 582 | 3501215 | 99 | |
| U95030.1 | - | 3 - 4 | 393 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alcam
Genomic Map