Assay Details
| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178925.3 | NP_849256.1 | 4 - 5 | 652 | 86 | |
| GenBank mRNA | AK032127.1 | - | 4 - 5 | 694 | 86 | |
| AK084698.1 | - | 4 - 5 | 646 | 86 | ||
| AK153942.1 | - | 4 - 5 | 466 | 86 | ||
| AK169841.1 | - | 4 - 5 | 455 | 86 | ||
| BC125628.1 | - | 4 - 5 | 652 | 40130143 | 86 | |
| BC125630.1 | - | 4 - 5 | 652 | 40130145 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK153942.1, AK169841.1 transcripts.
Set Membership: |
|

Nsun3
Genomic Map