Assay Details
| Gene Symbol: Arhgap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175535.3 | NP_780744.2 | 11 - 12 | 1462 | 113 | |
| GenBank mRNA | AK129345.1 | - | 8 - 9 | 951 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap20
Genomic Map