Assay Details
| Gene Symbol: Arhgap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175535.3 | NP_780744.2 | 12 - 13 | 1576 | 111 | |
| GenBank mRNA | AK129345.1 | - | 9 - 10 | 1065 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap20
Genomic Map