Assay Details
| Gene Symbol: Arhgap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175535.3 | NP_780744.2 | 8 - 9 | 935 | 91 | |
| GenBank mRNA | AK040756.1 | - | 8 - 9 | 935 | 91 | |
| AK129345.1 | - | 5 - 6 | 424 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap20
Genomic Map