Assay Details
| Gene Symbol: Arhgap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175535.3 | NP_780744.2 | 10 - 11 | 1297 | 88 | |
| GenBank mRNA | AK129345.1 | - | 7 - 8 | 786 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap20
Genomic Map