Assay Details
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010140.3 | NP_034270.1 | 2 - 3 | 266 | 86 | |
GenBank mRNA | AK041935.1 | - | 2 - 3 | 267 | 86 | |
AK045219.1 | - | 2 - 3 | 267 | 86 | ||
AK084921.1 | - | 2 - 3 | 246 | 86 | ||
AK132335.1 | - | 2 - 3 | 215 | 86 | ||
BC060164.1 | - | 2 - 3 | 243 | 86 | ||
BC093483.1 | - | 267 | 86 | |||
BC138641.1 | - | 2 - 3 | 189 | 8861661 | 86 | |
BC138643.1 | - | 2 - 3 | 189 | 8861663 | 86 | |
BC145349.1 | - | 2 - 3 | 189 | 9053905 | 86 | |
M68513.1 | - | 2 - 3 | 244 | 86 | ||
M68515.1 | - | 2 - 3 | 244 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
