Assay Details
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010140.3 | NP_034270.1 | 6 - 7 | 1543 | 101 | |
GenBank mRNA | AK041935.1 | - | 6 - 7 | 1544 | 101 | |
AK045219.1 | - | 6 - 7 | 1544 | 101 | ||
AK084921.1 | - | 6 - 7 | 1523 | 101 | ||
AK132335.1 | - | 6 - 7 | 1492 | 101 | ||
BC053479.1 | - | 2 - 3 | 435 | 101 | ||
BC060164.1 | - | 6 - 7 | 1520 | 101 | ||
BC093483.1 | - | 1544 | 101 | |||
BC138641.1 | - | 6 - 7 | 1466 | 8861661 | 101 | |
BC138643.1 | - | 6 - 7 | 1466 | 8861663 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
