Assay Details
Gene Symbol: Sept12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027669.3 | NP_081945.1 | 5 - 6 | 640 | 72 | |
NM_029374.1 | NP_083650.1 | 5 - 6 | 640 | 72 | ||
GenBank mRNA | AK006458.1 | - | 5 - 6 | 578 | 72 | |
AK016798.1 | - | 5 - 6 | 568 | 72 | ||
AK077091.1 | - | 5 - 6 | 641 | 72 | ||
BC115618.1 | - | 5 - 6 | 551 | 40102763 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
