Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900022278.mm.1 |
NM_027150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022278.mm.1 |
NM_027150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022278.mm.1 |
TC0900002698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022279.mm.1 |
NM_027150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022279.mm.1 |
NM_027150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022279.mm.1 |
TC0900002698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022277.mm.1 |
NM_027150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022277.mm.1 |
NM_027150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022277.mm.1 |
TC0900002698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022281.mm.1 |
NM_027150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022281.mm.1 |
NM_027150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022281.mm.1 |
TC0900002698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022290.mm.1 |
TC0900002698.mm.1 |
MTA TC |