Assay Details
| Gene Symbol: Anln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028390.3 | NP_082666.2 | 10 - 11 | 2066 | 71 | |
| GenBank mRNA | AK045703.1 | - | 7 - 8 | 71 | ||
| BC030489.1 | - | 3 - 4 | 408 | 5373071 | 71 | |
| BC032164.1 | - | 10 - 11 | 2084 | 5346703 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anln
Genomic Map