Assay Details
Gene Symbol: Anln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028390.3 | NP_082666.2 | 13 - 14 | 2505 | 93 | |
GenBank mRNA | AK045703.1 | - | 10 - 11 | 1469 | 93 | |
AK046102.1 | - | 2 - 3 | 291 | 93 | ||
BC030489.1 | - | 6 - 7 | 847 | 5373071 | 93 | |
BC032164.1 | - | 13 - 14 | 2523 | 5346703 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
