Assay Details
| Gene Symbol: Anln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028390.3 | NP_082666.2 | 5 - 6 | 1302 | 67 | |
| GenBank mRNA | AK013624.1 | - | 2 - 3 | 346 | 67 | |
| AK038403.1 | - | 2 - 3 | 377 | 67 | ||
| AK045703.1 | - | 2 - 3 | 266 | 67 | ||
| BC027141.1 | - | 5 - 6 | 1266 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anln
Genomic Map