Assay Details
| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145615.4 | NP_663590.3 | 3 - 4 | 445 | 87 | |
| GenBank mRNA | AK028118.1 | - | 3 - 4 | 446 | 87 | |
| AK032830.1 | - | 3 - 4 | 398 | 87 | ||
| AK167572.1 | - | 3 - 4 | 418 | 87 | ||
| AK168321.1 | - | 3 - 4 | 406 | 87 | ||
| AK218235.1 | - | 2 - 3 | 136 | 87 | ||
| BC003432.1 | - | 3 - 4 | 2646522 | 87 | ||
| BC096645.1 | - | 3 - 4 | 304 | 1245213 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etfa
Genomic Map