Assay Details
| Gene Symbol: Maats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081025.1 | NP_001074494.1 | 2 - 3 | 272 | 82 | |
| GenBank mRNA | AF503943.1 | - | 2 - 3 | 555 | 82 | |
| AK045121.1 | - | 2 - 3 | 224 | 82 | ||
| AK077038.1 | - | 2 - 3 | 258 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Maats1
Genomic Map