Assay Details
| Gene Symbol: Maats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081025.1 | NP_001074494.1 | 9 - 10 | 1277 | 86 | |
| GenBank mRNA | AK077038.1 | - | 9 - 10 | 1263 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maats1
Genomic Map