Assay Details
| Gene Symbol: Mpv17l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289561.1 | NP_001276490.1 | 3 - 4 | 489 | 168 | |
| NM_033564.3 | NP_291042.2 | 3 - 4 | 460 | 139 | ||
| GenBank mRNA | AF305634.1 | - | 3 - 4 | 430 | 139 | |
| BC094450.1 | - | 3 - 4 | 427 | 4973897 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpv17l
Genomic Map