Assay Details
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177326.3 | NP_796300.1 | 7 - 8 | 984 | 108 | |
GenBank mRNA | AK037275.1 | - | 7 - 8 | 985 | 109 | |
AY167030.1 | - | 6 - 7 | 710 | 108 | ||
BC063068.1 | - | 972 | 108 | |||
BC086650.1 | - | 7 - 8 | 915 | 6849008 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
