Assay Details
| Gene Symbol: Senp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177103.4 | NP_796077.2 | 6 - 7 | 2191 | 86 | |
| GenBank mRNA | AK028177.1 | - | 6 - 7 | 2194 | 86 | |
| AK043171.1 | - | 6 - 7 | 2076 | 86 | ||
| AY522918.1 | - | 6 - 7 | 2055 | 86 | ||
| BC067014.1 | - | 6 - 7 | 1958 | 6812169 | 86 | |
| BC080830.1 | - | 6 - 7 | 2075 | 30433573 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp5
Genomic Map