Assay Details
Gene Symbol: Iqch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030068.1 | NP_084344.1 | 4 - 5 | 409 | 69 | |
GenBank mRNA | AK019535.1 | - | 4 - 5 | 441 | 69 | |
AK029502.1 | - | 4 - 5 | 445 | 69 | ||
BC141047.1 | - | 4 - 5 | 409 | 9055659 | 69 | |
BC145231.1 | - | 4 - 5 | 409 | 9053783 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
